Anti-CD274 Mouse Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: Sino Biological
10084MM36F100 10084MM36F25
77581-654EA 92.7 USD
77581-654 77581-656
Anti-CD274 Mouse Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PD-L1/B7-H1/CD274 (rh PD-L1/B7-H1/CD274; Catalog#10084-H08H; NP_054862.1; Met1-Thr239) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.