Urine Collection Cups with Screw Cap
Supplier: SARSTEDT INC
75.9922.721 75.9922.744 75.9922.745
77589-076CS 343.46 USD
77589-076 77589-078 77589-080
Urine Collection Cups with Screw Cap
In order to meet the various requirements of hygienic collection, safe transportation and storage of diagnostic sample materials, this range offers many high-quality products for collecting urine, stool, saliva, CSF and tissue samples.
  • Material: PP
  • Flat base
  • Hygiene and convenience for patients and users
  • Highly transparent material
  • Odour-tight screw cap
  • Suitable for transport and storage of urine samples
  • With graduations and labeling area
Small volume urine containers for consistent and reliable samples.