Monovette® Urine Tubes with Stabilizer
Supplier: SARSTEDT INC
10.253 10.253.020
77587-332CS 372.91 USD
77587-332 77587-334
Monovette® Urine Tubes with Stabilizer
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
In order to meet the various requirements of hygienic collection, safe transportation and storage of diagnostic sample materials, this range offers many products for collecting urine, stool, saliva, CSF and tissue samples.
  • Material: PP
  • Preparation: Boric acid
  • Connection: Luer (m)
  • Closure type: Screw cap
  • Hygiene and protection for patients and users
  • Completely needle-free sample collection and processing
  • Developed for microbiological tests
  • Effective stabilization at room temperature for up to 48 hours
  • Primary container for transport as per ADR and IATA
The Urine Monovette® enables hygienic, needle-free sample collection from urine containers and urine drainage systems. Subsequently, a urine analysis with commercially available test strips can be carried out directly in the filled Urine Monovette®. If necessary, the Urine Monovette® is also suitable for collecting sediment in centrifuges and for further urine analysis in fully automated urine analysis systems. The Urine Monovette® can also be used as a transport container as per P650 of the ADR and IATA.
Light protection: No