InVivoSIM Anti-Claudin Human Antibody

Supplier: Bio X Cell
SIM0026-100MG SIM0026-1MG SIM0026-25MG SIM0026-50MG SIM0026-5MG
77580-760EA 7984.56 USD
77580-760 77580-762 77580-764 77580-766 77580-768
InVivoSIM Anti-Claudin Human Antibody
Antibodies
Zolbetuximab Biosimilar.

Type:
Antigen: Claudin
Clonality:
Clone:
Conjugation:
Epitope:
Host: Human
Isotype:
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen Symbol Claudin
Environmentally Preferable
Reactivity Human
Host Human
Isotype
Tested Applications Functional assays

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR