InVivoSIM Anti-PD-1 Human Antibody

Supplier: Bio X Cell
SIM0003-100MG SIM0003-1MG SIM0003-25MG SIM0003-50MG SIM0003-5MG
77580-530EA 7984.56 USD
77580-530 77580-532 77580-534 77580-536 77580-538
InVivoSIM Anti-PD-1 Human Antibody
Antibodies
Nivolumab Biosimilar.

Type:
Antigen: PD-1
Clonality:
Clone:
Conjugation:
Epitope:
Host: Human
Isotype: IgG4
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR