InVivoSIM Anti-EGFR Human Antibody

Supplier: Bio X Cell
SIM0002-100MG SIM0002-1MG SIM0002-25MG SIM0002-50MG SIM0002-5MG
77580-520EA 7984.56 USD
77580-520 77580-522 77580-524 77580-526 77580-528
InVivoSIM Anti-EGFR Human Antibody
Antibodies
Cetuximab Biosimilar.

Type:
Antigen: EGFR
Clonality:
Clone:
Conjugation:
Epitope:
Host: Human
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR