RecombiMAb Anti-PD-1 Mouse Recombinant Antibody [clone: RMP1-14-CP162]

Supplier: Bio X Cell
CP162-100MG CP162-1MG CP162-25MG CP162-50MG CP162-5MG
77580-370EA 6269.61 USD
77580-370 77580-372 77580-374 77580-376 77580-378
RecombiMAb Anti-PD-1 Mouse Recombinant Antibody [clone: RMP1-14-CP162]
Antibodies
Type: Primary
Antigen: PD-1
Clonality: Recombinant
Clone: RMP1-14-CP162
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR