RecombiMAb Anti-VEGFR-2 Mouse Recombinant Antibody [clone: DC101-CP132]

Supplier: Bio X Cell
CP132-100MG CP132-1MG CP132-25MG CP132-50MG CP132-5MG
77580-170EA 6269.61 USD
77580-170 77580-172 77580-174 77580-176 77580-178
RecombiMAb Anti-VEGFR-2 Mouse Recombinant Antibody [clone: DC101-CP132]
Antibodies
Type: Primary
Antigen: VEGFR-2
Clonality: Recombinant
Clone: DC101-CP132
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR