RecombiMAb Anti-CD8a Mouse Recombinant Antibody [clone: 2.43-CP128]

Supplier: Bio X Cell
CP128-100MG CP128-1MG CP128-25MG CP128-50MG CP128-5MG
77580-130EA 6269.61 USD
77580-130 77580-132 77580-134 77580-136 77580-138
RecombiMAb Anti-CD8a Mouse Recombinant Antibody [clone: 2.43-CP128]
Antibodies
Type: Primary
Antigen: CD8a
Clonality: Recombinant
Clone: 2.43-CP128
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR