InVivoMAb Anti-E protein Mouse Monoclonal Antibody [clone: POWV-63]

Supplier: Bio X Cell
BE0434-100MG BE0434-1MG BE0434-25MG BE0434-50MG BE0434-5MG
77579-880EA 4408.4 USD
77579-880 77579-882 77579-884 77579-886 77579-888
InVivoMAb Anti-E protein Mouse Monoclonal Antibody [clone: POWV-63]
Antibodies
Type: Primary
Antigen: E protein
Clonality: Monoclonal
Clone: POWV-63
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Powassan virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR