InVivoMAb Anti-CD2 Rat Monoclonal Antibody [clone: LO-CD2a]

Supplier: Bio X Cell
BE0406-100MG BE0406-1MG BE0406-25MG BE0406-50MG BE0406-5MG
77579-600EA 4408.4 USD
77579-600 77579-602 77579-604 77579-606 77579-608
InVivoMAb Anti-CD2 Rat Monoclonal Antibody [clone: LO-CD2a]
Antibodies
Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: LO-CD2a
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR