InVivoMAb Anti-IL-25 Mouse Monoclonal Antibody [clone: 2C3]

Supplier: Bio X Cell
BE0394-100MG BE0394-1MG BE0394-25MG BE0394-50MG BE0394-5MG
77579-480EA 4408.4 USD
77579-480 77579-482 77579-484 77579-486 77579-488
InVivoMAb Anti-IL-25 Mouse Monoclonal Antibody [clone: 2C3]
Antibodies
Type: Primary
Antigen: IL-25
Clonality: Monoclonal
Clone: 2C3
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR