InVivoMAb Anti-CD16.2 Armenian hamster Monoclonal Antibody [clone: 9E9]

Supplier: Bio X Cell
BE0378-100MG BE0378-1MG BE0378-25MG BE0378-50MG BE0378-5MG
77579-320EA 4408.4 USD
77579-320 77579-322 77579-324 77579-326 77579-328
InVivoMAb Anti-CD16.2 Armenian hamster Monoclonal Antibody [clone: 9E9]
Antibodies
Type: Primary
Antigen: CD16.2
Clonality: Monoclonal
Clone: 9E9
Conjugation:
Epitope:
Host: Armenian hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR