Urine Container with Integral Transfer Unit
Supplier: Sarstedt
75.562.400
77589-060CS 294.5 USD
77589-060
Urine Container with Integral Transfer Unit
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
To meet the various requirements of hygienic collection, safe transportation and storage of diagnostic sample materials, Sarstedt offers many high-quality products for collecting urine, stool, saliva, CSF and tissue samples.
  • Material: PP
  • Closure: Assembled
  • Odour-tight screw cap
  • 100% sterility guaranteed by the safety label
Hygiene and convenience for patients and users. Practical urine transfer to a V-Monovette® Urine from filling volumes of 20 ml with graduations and labelling area.
The Sarstedt V-Monovette® Urine evacuated system provides for hygienic urine collection, transfer, testing, and transport. V-Monovette® Urine evacuated tubes are available in a range of sizes from 4 to 10 ml with a round or conical base. Options with a boric acid stabilizer serve to maintain bacterial viability until analysis.
For a closed transfer into V-Monovette® Urine tubes, urine can be initially collected into a corresponding 100 ml urine cup with integrated sampling port. The sampling port needle is deeply recessed in the cap for improved safety and handling, and the cup is made from durable, highly transparent polypropylene. Alternatively, urine can be collected into any specimen cup and transferred into V-Monovette® Urine tubes via a transfer straw.