Transport Containers for Slides
Supplier: Sarstedt
86.972.150
77589-600CS 341.3 USD
77589-600
Transport Containers for Slides
Slide Mailers
Sarstedt offers sample transport systems and secondary containers for a wide variety of transport requirements.
  • Mailing container suitable for transport or storage of two slide plates (or four placed against each other)
  • Caps does not have ventilation
It offers standard and specialty transport systems for specimens.