Silicone Seals for Mini-Drums
Supplier: SAMPLING SYSTEMS LTD
SIL-6-FDA SIL-8-FDA SIL-10-FDA SIL-12-FDA SIL-15-FDA SIL-1-FDA SIL-2-FDA SIL-3-FDA SIL-5-FDA
77578-246EA 35.77 USD
77578-246 77578-248 77578-226 77578-228 77578-230 77578-232 77578-238 77578-240 77578-244
Silicone Seals for Mini-Drums
Seals Sealing Rings
Our O-ring Seals are sized to suit our range of Mini-Drums. These seals when used with toggle clamps will help seal the containers.
  • Easy to use
  • The silicone seal simply pushes on to the lid
These seals are made from high quality FDA compliant silicone. They are BSE/TSE free. Other materials are available on request. They can be fitted to a wide range of containers.