Inserts
Supplier: SARSTEDT INC
73.1051 73.1052
77589-024CS 154.09 USD
77589-024 77589-026
Inserts
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation
The high quality of blood samples collected with the S-Monovette® blood collection system is due to the gentle aspiration technique. It has been proven to significantly reduce hemolysis rates compared to vacuum systems. As a result, erythrocytes remain intact and blood collection does not have to be repeated as often.
  • For use in S-Monovette® and tubes with diameters of 13 and 15 mm
  • Ideal for low sample volume
  • Conical base shape for optimal serum or plasma yield
  • Material: PS