InVivoSIM Anti-HER2 Human Antibody

Supplier: Bio X Cell
SIM0020-100MG SIM0020-1MG SIM0020-25MG SIM0020-50MG SIM0020-5MG
77580-700EA 7984.56 USD
77580-700 77580-702 77580-704 77580-706 77580-708
InVivoSIM Anti-HER2 Human Antibody
Antibodies
Ustekinumab biosimilar.

Type:
Antigen: HER2
Clonality:
Clone:
Conjugation:
Epitope:
Host: Human
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR