InVivoPlus Anti-CD16/CD32 Rat Antibody [clone: 2.4G2]

Supplier: Bio X Cell
BP0307-100MG BP0307-25MG BP0307-50MG BP0307-5MG
77580-000EA 5965.76 USD
77580-000 77580-002 77580-004 77580-006
InVivoPlus Anti-CD16/CD32 Rat Antibody [clone: 2.4G2]
Antibodies
Type: Primary
Antigen: CD16/CD32
Clonality:
Clone: 2.4G2
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR