InVivoMAb Anti-pan-Glypican Human Monoclonal Antibody [clone: HS20]

Supplier: Bio X Cell
BE0412-100MG BE0412-1MG BE0412-25MG BE0412-50MG BE0412-5MG
77579-660EA 4408.4 USD
77579-660 77579-662 77579-664 77579-666 77579-668
InVivoMAb Anti-pan-Glypican Human Monoclonal Antibody [clone: HS20]
Antibodies
Type: Primary
Antigen: pan-Glypican
Clonality: Monoclonal
Clone: HS20
Conjugation:
Epitope:
Host: Human
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR