InVivoMAb Anti-GPC3 Mouse Monoclonal Antibody [clone: YP7]

Supplier: Bio X Cell
BE0404-100MG BE0404-1MG BE0404-25MG BE0404-50MG BE0404-5MG
77579-580EA 4408.4 USD
77579-580 77579-582 77579-584 77579-586 77579-588
InVivoMAb Anti-GPC3 Mouse Monoclonal Antibody [clone: YP7]
Antibodies
Type: Primary
Antigen: GPC3
Clonality: Monoclonal
Clone: YP7
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR