InVivoMAb Anti-CD96 Rat Monoclonal Antibody [clone: 6A6/CD96]

Supplier: Bio X Cell
BE0403-100MG BE0403-1MG BE0403-25MG BE0403-50MG BE0403-5MG
77579-570EA 4408.4 USD
77579-570 77579-572 77579-574 77579-576 77579-578
InVivoMAb Anti-CD96 Rat Monoclonal Antibody [clone: 6A6/CD96]
Antibodies
Type: Primary
Antigen: CD96
Clonality: Monoclonal
Clone: 6A6/CD96
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR