InVivoMAb Anti-RGMb Rat Monoclonal Antibody [clone: 307.9D1]

Supplier: Bio X Cell
BE0400-100MG BE0400-1MG BE0400-25MG BE0400-50MG BE0400-5MG
77579-540EA 4408.4 USD
77579-540 77579-542 77579-544 77579-546 77579-548
InVivoMAb Anti-RGMb Rat Monoclonal Antibody [clone: 307.9D1]
Antibodies
Type: Primary
Antigen: RGMb
Clonality: Monoclonal
Clone: 307.9D1
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR