InVivoMAb Anti-InVivoMAb anti-human/mouse GRP78 Mouse Monoclonal Antibody [clone: C38]

Supplier: Bio X Cell
BE0396-100MG BE0396-1MG BE0396-25MG BE0396-50MG BE0396-5MG
77579-500EA 4408.4 USD
77579-500 77579-502 77579-504 77579-506 77579-508
InVivoMAb Anti-InVivoMAb anti-human/mouse GRP78 Mouse Monoclonal Antibody [clone: C38]
Antibodies
Type: Primary
Antigen: InVivoMAb anti-human/mouse GRP78
Clonality: Monoclonal
Clone: C38
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR