InVivoMAb Anti-IL-12 Rat Monoclonal Antibody [clone: C18.2]

Supplier: Bio X Cell
BE0371-100MG BE0371-1MG BE0371-25MG BE0371-50MG BE0371-5MG
77579-250EA 4408.4 USD
77579-250 77579-252 77579-254 77579-256 77579-258
InVivoMAb Anti-IL-12 Rat Monoclonal Antibody [clone: C18.2]
Antibodies
Type: Primary
Antigen: IL-12
Clonality: Monoclonal
Clone: C18.2
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR