InVivoMAb Anti-CD71 (TfR1) Mouse Monoclonal Antibody [clone: B3/25]

Supplier: Bio X Cell
BE0367-100MG BE0367-1MG BE0367-25MG BE0367-50MG BE0367-5MG
77579-210EA 4408.4 USD
77579-210 77579-212 77579-214 77579-216 77579-218
InVivoMAb Anti-CD71 (TfR1) Mouse Monoclonal Antibody [clone: B3/25]
Antibodies
Type: Primary
Antigen: CD71 (TfR1)
Clonality: Monoclonal
Clone: B3/25
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR