InVivoMAb Anti-CD71 Mouse Monoclonal Antibody [clone: 5E9C11]

Supplier: Bio X Cell
BE0343-100MG BE0343-1MG BE0343-25MG BE0343-50MG BE0343-5MG
77578-980EA 4408.4 USD
77578-980 77578-982 77578-984 77578-986 77578-988
InVivoMAb Anti-CD71 Mouse Monoclonal Antibody [clone: 5E9C11]
Antibodies
Type: Primary
Antigen: CD71
Clonality: Monoclonal
Clone: 5E9C11
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR