InVivoMAb Anti-CD220 Mouse Monoclonal Antibody [clone: IR]

Supplier: Bio X Cell
BE0338-100MG BE0338-1MG BE0338-25MG BE0338-50MG BE0338-5MG
77578-930EA 4408.4 USD
77578-930 77578-932 77578-934 77578-936 77578-938
InVivoMAb Anti-CD220 Mouse Monoclonal Antibody [clone: IR]
Antibodies
Type: Primary
Antigen: CD220
Clonality: Monoclonal
Clone: IR
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR