InVivoMAb Anti-IL-27 Mouse Monoclonal Antibody [clone: MM27.7B1]

Supplier: Bio X Cell
BE0326-100MG BE0326-1MG BE0326-25MG BE0326-50MG BE0326-5MG
77578-810EA 4408.4 USD
77578-810 77578-812 77578-814 77578-816 77578-818
InVivoMAb Anti-IL-27 Mouse Monoclonal Antibody [clone: MM27.7B1]
Antibodies
Type: Primary
Antigen: IL-27
Clonality: Monoclonal
Clone: MM27.7B1
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR