InVivoMAb F(ab')2 Anti-IgG Hamster Monoclonal Antibody [clone: hamster]

Supplier: Bio X Cell
BE0091-FAB-50MG BE0091-FAB-10MG BE0091-FAB-25MG
77578-780EA 4302.31 USD
77578-780 77578-776 77578-778
InVivoMAb F(ab')2 Anti-IgG Hamster Monoclonal Antibody [clone: hamster]
Antibodies
Type:
Antigen: IgG
Clonality: Monoclonal
Clone: hamster
Conjugation:
Epitope:
Host: Hamster
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR