2-(5H-Chromeno[2,3-b]pyridin-7-yl)propanoic acid

Supplier: TCI America

Synonyms: Pranoprofen

P2778-1G P2778-5G
TCP2778-1GEA 210.12 USD
TCP2778-1G TCP2778-5G
2-(5H-Chromeno[2,3-b]pyridin-7-yl)propanoic acid
2-(5H-Chromeno[2,3-b]pyridin-7-yl)propanoic acid
Formula: C₁₅H₁₃NO₃
MW: 255.26 g/mol
MDL Number: MFCD00866111
CAS Number: 52549-17-4
UN: 2811

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR