NANOCOLOR® Photometric Test Kits
Supplier: CTL SCIENTIFIC SUPPLY CORP
91888 91895 91802 91805 918101 918126 918128 918131 918142 91816 918163 918172 91860 91862 91863 91865 91867 91875 91877 91878 91885 91820 91825 91830 91832 91834 91836 91844 91848 91851 91853
77578-662EA 209.97 USD
77578-662 77578-664 77578-606 77578-608 77578-638 77578-642 77578-636 77578-610 77578-630 77578-614 77578-616 77578-432 77578-640 77578-644 77578-648 77578-646 77578-650 77578-658 77578-652 77578-654 77578-656 77578-612 77578-618 77578-624 77578-626 77578-628 77578-634 77578-632 77578-660 77578-620 77578-622
NANOCOLOR® Photometric Test Kits
A range of analytical reagents for photometric water analysis in the NANOCOLOR® standard test version.
  • Excellent sensitivity and accuracy due to larger cuvette sizes
  • Lower costs
  • Easy dilutions for analysis of higher concentrations
Ready-to-use reagents that can be used for different sizes 10, 20 and 50 mm of cuvettes. Tests that are particularly suitable for the NANOCOLOR® photometers but can be used with other photometers.
Ordering information: For details of the full range of all NANOCOLOR® test kits please contact Avantor.