Amiodarone hydrochloride

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Butyl-3-benzofuranyl 4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl ketone hydrochloride)(2-Butylbenzofuran-3-yl)(4-(2-(diethylamino)ethoxy)-3,5-diiodophenyl)methanone hydrochlorideAmiodarone HCl

457980010 457980050
TS457980010EA 21.9 USD
TS457980010 TS457980050
Amiodarone hydrochloride
Amiodarone hydrochloride
Formula: C₂₅H₂₉I₂NO₃
MW: 681.78 g/mol
Melting Pt: 156 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00069204
CAS Number: 19774-82-4

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 %
Sulfated ash ≤0.1 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR