Imidazo[1,2-b]pyridazine 98%

Supplier: Thermo Scientific Chemicals
Warning

455660010 455660050
TS455660010EA 25 USD
TS455660010 TS455660050
Imidazo[1,2-b]pyridazine 98%
Imidazo[1,2-b]pyridazine
Formula: C₆H₅N₃
MW: 119.12 g/mol
Melting Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD07782103
CAS Number: 766-55-2

Order Now

Specification Test Results

Appearance Yellow to light grey Solid
Infrared spectrum Conforms
Melting point 50 to 57 °C
Water ≤3 %
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR