Boron nitride 98%

Supplier: Thermo Scientific Chemicals

441800500 441802500
TS441800500EA 120 USD
TS441800500 TS441802500
Boron nitride 98%
Boron nitride
Formula: BN
Melting Pt: 2700…3000 °C
Storage Temperature: Ambient
MDL Number: MFCD00011317
CAS Number: 10043-11-5

Order Now

Specification Test Results

Appearance White to off-white Powder
Assay ≥97.5 %
Particle size 0.17 to 5 µm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR