Reinecke's salt ACS

Supplier: Thermo Scientific Chemicals
Warning

423330250 423331000 423335000
TS423330250EA 66 USD
TS423330250 TS423331000 TS423335000
Reinecke's salt ACS
Reinecke's salt
Formula: C₄H₁₀CrN₇S₄
Melting Pt: 270 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00151654
CAS Number: 13573-16-5
EINECS: 237-003-3

Order Now

Specification Test Results

Appearance Red Crystalline powder
Infrared spectrum Conforms
Gravimetry ≥93.0 %
Sensitivity passes test
Insoluble matter ≤0.05 % (in dilute HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR