2-Isopropyl-N,2,3-trimethylbutyramide

Supplier: Thermo Scientific Chemicals
Warning

421930050 421930250
TS421930050EA 26 USD
TS421930050 TS421930250
2-Isopropyl-N,2,3-trimethylbutyramide
2-Isopropyl-N,2,3-trimethylbutyramide
Formula: C₁₀H₂₁NO
Melting Pt: 58…61 °C
Storage Temperature: Ambient
MDL Number: MFCD00130070
CAS Number: 51115-67-4

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Melting point 60 to 65 °C
Water ≤0.5 %
GC ≥99.4 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR