Tin ACS 20 mesh
Supplier: Thermo Scientific Chemicals
420981000
420985000
TS420981000EA
78
USD
TS420981000
TS420985000
Tin ACS 20 mesh
Tin
Formula:
Sn Boiling Pt: 2270 °C (760 mmHg) Melting Pt: 231.9 °C Storage Temperature: Ambient |
MDL Number:
MFCD00133862 CAS Number: 7440-31-5 EINECS: 231-141-8 |
Specification Test Results
Appearance | Grey Metallic granular powder |
Titration Complexometric | ≥99.5 % |
Antimony (Sb) | ≤200 ppm |
Arsenic (As) | ≤1 ppm |
Copper (Cu) | ≤50 ppm |
Iron (Fe) | ≤100 ppm |
Lead (Pb) | ≤50 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...