Silica gel, Pore size: 150 Å 0.075-0.250 mm for chromatography
Supplier: Thermo Scientific Chemicals
419280010
419280050
419282500
TS419280010EA
252
USD
TS419280010
TS419280050
TS419282500
Silica gel, Pore size: 150 Å 0.075-0.250 mm for chromatography
Silica gel
Formula:
SiO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00011232 CAS Number: 63231-67-4 REACH: 01-2119379499-16 |
Specification Test Results
Appearance | White Granular powder |
Particle size | ≤10.0 % < 200 Mesh |
Specific surface area | 275 to 375 m²/g (BET) |
Pore volume | 0.95 to 1.3 ml/g (N2) |
Volatiles | ≤8.0 % (160°C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...