Charcoal activated, NORIT® CN 1 for decolorizing

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Activated carbonCarbon activatedActivated charcoal

404042500 404045000
TS404045000EA 61.3 USD
TS404045000 TS404042500
Charcoal activated, NORIT® CN 1 for decolorizing
Charcoal activated
Formula: C
MW: 12.01 g/mol
Melting Pt: 3652 °C
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
UN: 1362
ADR: 4.2,III
REACH: 01-2119488894-16
Merck Index: 13,01818

Order Now

Specification Test Results

Appearance Black Powder
Loss on drying ≤15 %
Methylene blue adsorption ≥25.0 g/100 g
pH 5.5 to 8
Phosphate (PO4) ≤3.5 % (by acid extraction)
Calcium (Ca) ≤200 ppm (by acid extraction)
Iron (Fe) ≤150 ppm (by acid extraction)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR