3-Methylthiophene-2-carbonyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

380400020 380400100
TS380400020EA 29 USD
TS380400020 TS380400100
3-Methylthiophene-2-carbonyl chloride 98%
3-Methylthiophene-2-carbonyl chloride
Formula: C₆H₅ClOS
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00130091
CAS Number: 61341-26-2
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5830 to 1.5850 (20 °C, 589 nm)
Titration RCOCl ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR