4-Fluoro-DL-phenylglycine 98+%
Supplier: Thermo Scientific Chemicals
Synonyms:
4-Fluoro-DL-α-phenylglycine, 4-Fluorophenylglycine, H-DL-Phg(4-F)-OH
367930010
367930050
TS367930010EA
27
USD
TS367930010
TS367930050
4-Fluoro-DL-phenylglycine 98+%
4-Fluoro-DL-phenylglycine
Formula:
C₈H₈FNO₂ MW: 169.155 g/mol Melting Pt: 300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00066444 CAS Number: 7292-73-1 |
Specification Test Results
Appearance | Light yellow Powder |
Infrared spectrum | Conforms |
HPLC | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...