Boc-cycloleucine 98%
Supplier: Thermo Scientific Chemicals
367340010
367340050
TS367340010EA
37
USD
TS367340010
TS367340050
Boc-cycloleucine 98%
Boc-cycloleucine
Formula:
C₁₁H₁₉NO₄ Melting Pt: 130…135 °C Storage Temperature: Ambient |
MDL Number:
MFCD01076126 CAS Number: 35264-09-6 |
Specification Test Results
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 130 to 135 °C |
Titration with NaOH | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...