Silver 99.9%, foil 25x25 mm, Thickness 1.0 mm , 1ea = 6.6gr

Supplier: Thermo Scientific Chemicals

317750010 317750040
TS317750010EA 130 USD
TS317750010 TS317750040
Silver 99.9%, foil 25x25 mm, Thickness 1.0 mm , 1ea = 6.6gr
Silver
Formula: Ag
Boiling Pt: 2212 °C (1013 hPa)
Melting Pt: 961 °C
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Order Now

Specification Test Results

Appearance Silver white Foil
Total trace metal impurities ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR