Silica gel, Pore size: 40 Å 0.200-0.500 mm for chromatography

Supplier: Thermo Scientific Chemicals

241670010 241672500
TS241670010EA 160 USD
TS241670010 TS241672500
Silica gel, Pore size: 40 Å 0.200-0.500 mm for chromatography
Silica gel
Formula: SiO₂
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 63231-67-4
REACH: 01-2119379499-16

Order Now

Specification Test Results

Appearance White Crystals
Particle size ≤5 % (Oversize)
Specific gravity 0.54 to 0.6 kg/l
pH 5.5 to 7 (10 % suspension in water)
Residual water 2 to 4 % (160 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR