Silica gel, Pore size: 40 Å 0.060-0.200 mm for chromatography
Supplier: Thermo Scientific Chemicals
241660010
241660050
TS241660010EA
154
USD
TS241660010
TS241660050
Silica gel, Pore size: 40 Å 0.060-0.200 mm for chromatography
Silica gel
Formula:
SiO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00011232 CAS Number: 63231-67-4 REACH: 01-2119379499-16 |
Specification Test Results
Appearance | White Powder |
Particle size | ≤10 % (Oversize) |
Specific surface area | ≥500 m²/g |
pH | 5.5 to 8.0 (5 % suspension in water) |
Residual water | 2 to 4 % (160°C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...