Copper 99%, powder max 106 µm (-140 mesh)

Supplier: Thermo Scientific Chemicals
Warning

196570025 196570250 196575000
TS196570025EA 197 USD
TS196570025 TS196570250 TS196575000
Copper 99%, powder max 106 µm (-140 mesh)
Copper
Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
UN: 3089
ADR: 4.1 III
Merck Index: 13,02545

Order Now

Specification Test Results

Appearance Red to red-brown Powder
Assay ≥98.5 % (Cu)
Particle size (Maximum) 106 µm
Apparent density 1.35 g/ml
Average particle size 25 µm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR