Indium 99.999% (trace metals basis), powder
Supplier: Thermo Scientific Chemicals
|
Warning
|
193180050
193180250
193181000
TS193180050EA
142
USD
TS193180050
TS193180250
TS193181000
Indium 99.999% (trace metals basis), powder
Indium
Formula:
In Boiling Pt: 2000 °C (760 mmHg) Melting Pt: 156 °C Density: 7.3 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00134048 CAS Number: 7440-74-6 EINECS: 231-180-0 UN: 3089 ADR: 4.1,II |
Specification Test Results
Appearance | Grey Powder |
Total trace metal impurities | ≤0.001 % |
Anticaking agent | 2 % (MgO) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...