Cobalt 99.995% (trace metals basis), powder and/or granules

Supplier: Thermo Scientific Chemicals
Danger

192850050 192850250
TS192850050EA 86 USD
TS192850050 TS192850250
Cobalt 99.995% (trace metals basis), powder and/or granules
Cobalt
Formula: Co
Boiling Pt: 2870 °C (760 mmHg)
Melting Pt: 1495 °C
Storage Temperature: Ambient
MDL Number: MFCD00010935
CAS Number: 7440-48-4

Order Now

Specification Test Results

Appearance Grey Powder and/or granules
Total trace metal impurities ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR