S-Monovette® Venous Blood Collection Tubes
Supplier: SARSTEDT INC
04.1901.100 04.1902.100 04.1903.100 04.1904.100 04.1905.001 04.1906.100 04.1907.100 04.1913.100 04.1913.110 03.1068.100 03.1397.100 03.1524.001 03.1628.100 03.1631.100 04.1900.100 06.1660.100 06.1663.100 06.1664.100 06.1665.100 06.1666.100 06.1667.001 06.1668.100 06.1669.100 04.1926.001 04.1928.100 04.1929.100 04.1931.100 04.1933.100 04.1934.100 04.1935.001 04.1936.100 04.1940.100 04.1943.100 05.1165.100 05.1167.100 05.1553.100 05.1557.100 05.1729.001 01.1601.100 01.1602.001 01.1604.100 01.1604.400 01.1605.100 01.1613.100 01.1624.001 01.1728.001 04.1914.100 04.1915.100 04.1917.100 04.1918.100 04.1919.100 04.1920.100 04.1922.100 04.1923.001 04.1924.100 04.1925.001 04.1951.100 04.1955.100 05.1073.100 05.1081.100
77587-202CS 210.85 USD
77587-202 77587-204 77587-206 77587-208 77587-210 77587-212 77587-214 77587-216 77587-218 77587-220 77587-182 77587-184 77587-190 77587-192 77587-194 77587-196 77587-198 77587-200 77587-302 77587-304 77587-306 77587-308 77587-310 77587-312 77587-314 77587-316 77587-242 77587-244 77587-246 77587-248 77587-250 77587-252 77587-254 77587-256 77587-258 77587-260 77587-292 77587-294 77587-296 77587-298 77587-300 77587-162 77587-164 77587-166 77587-168 77587-170 77587-172 77587-174 77587-176 77587-178 77587-180 77587-222 77587-224 77587-226 77587-228 77587-230 77587-232 77587-234 77587-236 77587-238 77587-240 77587-262 77587-264 77587-266 77587-268 77587-270 77587-274 77587-278
S-Monovette® Venous Blood Collection Tubes
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
The high quality of blood samples collected with the S-Monovette® blood collection system is due to the gentle aspiration technique. It has been proven to significantly reduce hemolysis rates compared to vacuum systems. As a result, erythrocytes remain intact and blood collection does not have to be repeated as often.
  • Full suite of preparations for needed testing
  • Available in large and small volumes starting from 1.1 ml
  • Centrifugation: min. 2000×g to max. 4000×g
  • The screw cap makes the S-Monovette® easy to open, thereby minimizing the aerosol effect
Due to the gentle aspiration technique, the S-Monovette® blood collection system is suitable for all vein conditions, from young to old. It offers optimal sample quality and the best results thanks to the revolutionary blood collection system.
This also includes a well-designed, environmentally friendly packaging concept to reduce waste and a ready-to-use system for the needles. All needles can be used straight out of the packaging and allow users to work quickly - there is no assembly required as the needles are available exclusively with integrated holders.
In addition to a premium blood collection system, we support our customers in their everyday work with a pre-analytical consulting group. A global team of experts is available for any pre-analytical questions, areas needing improvement, or product training.